Learn More
Tocris Bioscience™ 5J 4
CRAC channel blocker; anti-inflammatory
Brand:  Tocris Bioscience™ 5490/10
141.68 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18766597
  • 180.00 € / 10mg
Explore more special offers
Description
Description
5J 4 is a calcium release-activated calcium (CRAC) channel blocker. Blocks store-operated calcium entry (SOCE) in HeLa-O+S cells and Th17 cells in vitro. Ameliorates symptoms and reduces CNS infiltration by mononuclear cells in a mouse model of experimental autoimmune encephalomyelitis (EAE) in vivo.
Specifications
Specifications
N-[[(6-Hydroxy-1-naphthalenyl)amino]thioxomethyl]-2-furancarboxamide
10 mg
C16H12N2O3S
>98%
827001-82-1
Other Calcium Channel Blockers
312.34