missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 5J 4
CRAC channel blocker; anti-inflammatory
180.00 € - 695.00 €
Especificaciones
Chemical Name or Material | N-[[(6-Hydroxy-1-naphthalenyl)amino]thioxomethyl]-2-furancarboxamide |
---|---|
CAS | 827001-82-1 |
Target | Other Calcium Channel Blockers |
Molecular Formula | C16H12N2O3S |
Formula Weight | 312.34 |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
18766597
|
Tocris Bioscience™
5490/10 |
10 mg |
180.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18715688
|
Tocris Bioscience™
5490/50 |
50 mg |
695.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
Calcium release-activated calcium (CRAC) channel blocker. Blocks store-operated calcium entry (SOCE) in HeLa-O+S cells and Th17 cells in vitro. Ameliorates symptoms and reduces CNS infiltration by mononuclear cells in a mouse model of experimental autoimmune encephalomyelitis (EAE) in vivo.Especificaciones
N-[[(6-Hydroxy-1-naphthalenyl)amino]thioxomethyl]-2-furancarboxamide | |
Other Calcium Channel Blockers | |
312.34 |
827001-82-1 | |
C16H12N2O3S | |
>98% |
Seguridad y manipulación
Recommended Storage : Store at -20°C