missing translation for 'onlineSavingsMsg'
Learn More

Heidolph Instruments Heating Bath Liquid

For use with heated baths

Brand:  Heidolph Instruments 515-31000-00

Product Code. 11711699

  • 472.00 € / 5 L
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Heating bath liquid up to 170 °C (5 L)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 L
240°C
Rotary Evaporator
115/230 VAC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.