missing translation for 'onlineSavingsMsg'
Learn More

(5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone, 98%, Thermo Scientific Chemicals

Product Code. 15497318
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 100516-54-9
Melting Point 205°C to 207°C
Molecular Formula C24H21NO4
MDL Number MFCD00074958
Quantity 1 g
Synonym 2r,3s-benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate, benzyl 2r,3s---6-oxo-2,3-diphenyl-4-morpholinecarboxylate, benzyl 2r,3s-6-oxo-2,3-diphenylmorpholine-4-carboxylate, 2r,3s-n-cbz-6-oxo-2,3-diphenylmorpholine, 2r,3s---n-z-6-oxo-2,3-diphenylmorpholine, benzyl 2r,3s-6-oxo-2,3-diphenyl-4-morpholinecarboxylate, pubchem18045, 2r,3s-6-oxo-2,3-diphenyl-4-morpholinecarboxylate
InChI Key HECRUWTZAMPQOS-UHFFFAOYNA-N
SMILES O=C(OCC1=CC=CC=C1)N1CC(=O)OC(C1C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
Molecular Weight (g/mol) 387.44
PubChem CID 981238
Formula Weight 387.43
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'