missing translation for 'onlineSavingsMsg'
Learn More

5X Kinase Buffer A

5X Kinase Buffer A

200.00 € - 742.00 €

Specifications

Concentration 5X
For Use With (Application) Pharma & Biopharma; Protein Assays and Analysis
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Type Kinase Buffer A
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10013013
View Documents
Invitrogen
PV3189
4 mL
200.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11300972
View Documents
Invitrogen
PV6135
100 ml
742.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 5X Kinase Buffer A is a concentrated general kinase assay buffer that can be used with a variety of assay technologies such as, LanthaScreen™, Adapta™, and Z’-LYTE™ assays.

Ready to use and validated with LanthaScreen™, Adapta™, and Z’LYTE™ assays

  • Optimized 5X concentrated formulation
  • Simplified handling with room temperature storage

    Easily Screen for Kinase Targets using an Invitrogen™ Platform
    For more information on the kinase assays that can be used with our 5X Kinase Buffer A see the following links:
    LanthaScreen™ Eu Binding Assay
    LanthaScreen™ Activity Assay
    Adapta™ Universal Kinase Assay
    Z´-LYTE™ Kinase Assay Kits

    Save Freezer Space
    The 5X Kinase Buffer can be stored at room temperature, saving you time and precious freezer space.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
  • Specifications

    Specifications

    5X
    Approved for shipment at Room Temperature or on Wet or Dry Ice
    Liquid
    Pharma & Biopharma; Protein Assays and Analysis
    Kinase Buffer A
    Videos
    SDS
    Documents

    Documents

    Product Certifications

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.