missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ II Green PCR Buffer (5X)

Invitrogen 5X Platinum II Green PCR Buffer for fast and easy PCR

Brand:  Invitrogen™ 14966123

Product Code. 16697075

  • 119.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 4 x 1.25 ml 5X Platinum II Green PCR buffer
This item is not returnable. View return policy

Description

Description

Invitrogen Platinum II Green PCR Buffer is an innovative buffer optimized for use with Platinum II Taq Hot-Start DNA Polymerase. Due to its unique composition, the annealing temperature is 60°C for most primer pairs designed following the general design rules. Isostabilizing molecules in the buffer increase primer–template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize annealing temperature for each primer pair.

Platinum II Green PCR Buffer provides 1.5 mM MgCl2 in the final reaction and is supplemented with a density reagent and two tracking dyes for direct loading of PCR products on gels.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5X Platinum II Green PCR buffer, 4 x 1.25 mL

Store at -20°C in a non-frost-free freezer.

Platinum II Taq Hot-Start DNA Polymerase
PCR Buffer
5X
Tube
Fast
5 mL
4 x 1.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.