missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(2-furyl)nicotinic acid, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Formula Weight | 189.17 |
Percent Purity | 97% |
Chemical Name or Material | 6-(2-Furyl)nicotinic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction