missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-1-tetralone, 97%, Thermo Scientific Chemicals

Product Code. 11934871
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13575-75-2
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00134100
InChI Key YNNJHKOXXBIJKK-UHFFFAOYSA-N
Synonym 6,7-dimethoxy-1-tetralone, 6,7-dimethoxy-3,4-dihydronaphthalen-1 2h-one, 6,7-dimethoxytetralin-1-one, 6,7-dimethoxy-3,4-dihydro-1 2h-naphthalenone, 1 2h-naphthalenone, 3,4-dihydro-6,7-dimethoxy, 3,4-dihydro-6,7-dimethoxynaphthalen-1 2h-one, 6,7-dimethoxy-2,3,4-trihydronaphthalen-1-one, 7-dimethoxy, 4-dihydro-6, zlchem 843
PubChem CID 266816
IUPAC Name 6,7-dimethoxy-3,4-dihydro-2H-naphthalen-1-one
SMILES COC1=CC2=C(C=C1OC)C(=O)CCC2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 95.0°C to 98.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 206.24
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 6, 7-Dimethoxy-1-tetralone
Show More Show Less
compliance-icons
Product Identifier
  • 6,7-Dimethoxy-1-tetralone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.