missing translation for 'onlineSavingsMsg'
Learn More

6-Acetyl-2H-1,4-benzoxazin-3(4H)-one, 98%, Thermo Scientific™

Product Code. 11337219
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26518-71-8
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD01321313
InChI Key BKJFWHFUERNXLJ-UHFFFAOYSA-N
PubChem CID 3641956
IUPAC Name 6-acetyl-4H-1,4-benzoxazin-3-one
SMILES CC(=O)C1=CC2=C(C=C1)OCC(=O)N2

Specifications

Melting Point 193°C to 199°C
Quantity 5 g
Synonym 6-acetyl-2h-1,4-benzoxazin-3 4h-one, 6-acetyl-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 6-acetyl, 6-acetyl-2,4-dihydro-1,4-benzoxazin-3-one, acmc-1co8k, 6-acetyl-4h-benzo 1,4 oxazin-3-one, 6-acetyl-2h-1,4-benzoxazine-3 4h-one, 2h-1,4-benzoxazin-3 4h-one,6-acetyl, 6-acetyl-2h-1,4-benzoxazin-3 4 h-one
Formula Weight 191.19
Percent Purity 98%
Chemical Name or Material 6-Acetyl-2H-1,4-benzoxazin-3(4H)-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'