missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-3-methyl-1,3-benzoxazol-2(3H)-one, 90%, Thermo Scientific Chemicals

Product Code. 10853001
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99584-10-8
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.16
MDL Number MFCD07801206
InChI Key FPNLXQSOWBNXCN-UHFFFAOYSA-N
Synonym 6-amino-3-methyl-3h-benzooxazol-2-one, 6-amino-3-methyl-1,3-benzoxazol-2 3h-one, 6-amino-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one, 6-amino-3-methyl-3-hydrobenzoxazol-2-one, 6-amino-3-methylbenzoxazol-2-one, 2 3h-benzoxazolone, 6-amino-3-methyl, 6-amino-3-methylbenzo d oxazol-2 3h-one
PubChem CID 6486006
IUPAC Name 6-amino-3-methyl-1,3-benzoxazol-2-one
SMILES CN1C(=O)OC2=CC(N)=CC=C12

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 148°C to 151°C
Color Tan
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 164.17
Percent Purity 90%
Physical Form Powder
Chemical Name or Material 6-Amino-3-methyl-1, 3-benzoxazol-2(3H)-one
Show More Show Less
compliance-icons
Product Identifier
  • 6-Amino-3-methyl-1,3-benzoxazol-2(3H)-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'