Learn More
6-Amino-5-azacytidine, MedChemExpress
MedChemExpress 6-Amino-5-azacytidine inhibits the growth of bacteria E. coli.
Brand:  Medchem Express HY-111643
Encompass_Preferred
Product Code. 16454508
  • 464.40 € / 10mg
Explore more special offers
Description
Description
6-Amino-5-azacytidine inhibits the growth of bacteria E. coli.
Specifications
Specifications
6-Amino-5-azacytidine
White
C8H13N5O5
H2O : 14.71 mg/mL (56.75 mM; Need ultrasonic)
259.22
95.0%
Solid
105331-00-8
Research
10 mg
O=C1N([C@H]2[C@H](O)[C@H](O)[C@@H](CO)O2)C(N)=NC(N)=N1
259.22
Research
COVID-19-immunoregulation