missing translation for 'onlineSavingsMsg'
Learn More

6-Benzoyl-3-(alpha-cyanobenzyl)-2(1H)-quinoxalinone, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1858242-02-0
Molecular Formula C23H15N3O2
Molecular Weight (g/mol) 365.392
MDL Number MFCD22988965
InChI Key JAHIUCLUJXQZRA-UHFFFAOYSA-N
Synonym 2-7-benzoyl-3-oxo-4h-quinoxalin-2-yl-2-phenylacetonitrile, 6-benzoyl-3-alpha-cyanobenzyl-2 1h-quinoxalinone
PubChem CID 121235645
IUPAC Name 2-(7-benzoyl-3-oxo-4H-quinoxalin-2-yl)-2-phenylacetonitrile
SMILES C1=CC=C(C=C1)C(C#N)C2=NC3=C(C=CC(=C3)C(=O)C4=CC=CC=C4)NC2=O
View More Specs

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1858242-02-0
365.392
JAHIUCLUJXQZRA-UHFFFAOYSA-N
121235645
C1=CC=C(C=C1)C(C#N)C2=NC3=C(C=CC(=C3)C(=O)C4=CC=CC=C4)NC2=O
C23H15N3O2
MFCD22988965
2-7-benzoyl-3-oxo-4h-quinoxalin-2-yl-2-phenylacetonitrile, 6-benzoyl-3-alpha-cyanobenzyl-2 1h-quinoxalinone
2-(7-benzoyl-3-oxo-4H-quinoxalin-2-yl)-2-phenylacetonitrile

Specifications

1858242-02-0
MFCD22988965
2-7-benzoyl-3-oxo-4h-quinoxalin-2-yl-2-phenylacetonitrile, 6-benzoyl-3-alpha-cyanobenzyl-2 1h-quinoxalinone
C1=CC=C(C=C1)C(C#N)C2=NC3=C(C=CC(=C3)C(=O)C4=CC=CC=C4)NC2=O
365.392
365.39
6-Benzoyl-3-(alpha-cyanobenzyl)-2(1H)-quinoxalinone
C23H15N3O2
UN3439
JAHIUCLUJXQZRA-UHFFFAOYSA-N
2-(7-benzoyl-3-oxo-4H-quinoxalin-2-yl)-2-phenylacetonitrile
121235645
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.