Learn More
6-(Benzylamino)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
124.00 € - 298.00 €
Chemical Identifiers
CAS | 1073354-27-4 |
---|---|
Molecular Formula | C18H23BN2O2 |
Molecular Weight (g/mol) | 310.204 |
MDL Number | MFCD06798270 |
InChI Key | BWIHFPRFKBKWMU-UHFFFAOYSA-N |
Synonym | 6-benzylamino pyridine-3-boronic acid pinacol ester, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, n-benzyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, amtb753, 6-benzylamino pyridine-3-boronicacidpinacolester, 2-benzylamino pyridine-5-boronic acid pinacol ester, n-phenylmethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinamine, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-pyridinamine, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-amine, 2-pyridinamine, n-phenylmethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl |
PubChem CID | 44755164 |
IUPAC Name | N-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)NCC3=CC=CC=C3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15417695
|
Thermo Scientific Alfa Aesar
H27463.MD |
250 mg |
124.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377329
|
Thermo Scientific Alfa Aesar
H27463.03 |
1 g |
298.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
1073354-27-4 | |
310.204 | |
BWIHFPRFKBKWMU-UHFFFAOYSA-N | |
44755164 | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)NCC3=CC=CC=C3 |
C18H23BN2O2 | |
MFCD06798270 | |
6-benzylamino pyridine-3-boronic acid pinacol ester, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, n-benzyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, amtb753, 6-benzylamino pyridine-3-boronicacidpinacolester, 2-benzylamino pyridine-5-boronic acid pinacol ester, n-phenylmethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinamine, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-pyridinamine, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-amine, 2-pyridinamine, n-phenylmethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
N-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine |
Specifications
1073354-27-4 | |
C18H23BN2O2 | |
6-benzylamino pyridine-3-boronic acid pinacol ester, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, n-benzyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, amtb753, 6-benzylamino pyridine-3-boronicacidpinacolester, 2-benzylamino pyridine-5-boronic acid pinacol ester, n-phenylmethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinamine, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-pyridinamine, n-benzyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridin-2-amine, 2-pyridinamine, n-phenylmethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
BWIHFPRFKBKWMU-UHFFFAOYSA-N | |
N-benzyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine | |
44755164 | |
95% |
135°C to 138°C | |
MFCD06798270 | |
Insoluble in water. | |
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)NCC3=CC=CC=C3 | |
310.204 | |
310.2 | |
6-(Benzylamino)pyridine-3-boronic acid pinacol ester |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.