missing translation for 'onlineSavingsMsg'
Learn More
6-Benzylaminopurine, 99%, Thermo Scientific Chemicals Product Code.: 10637542

6-Benzylaminopurine, 99%, Thermo Scientific Chemicals

Product Code. 10637542
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10637542

Brand: Thermo Scientific Acros 226410010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214-39-7
Molecular Formula C12H11N5
Molecular Weight (g/mol) 225.26
MDL Number MFCD00005572
InChI Key NWBJYWHLCVSVIJ-UHFFFAOYSA-N
Synonym 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
PubChem CID 62389
ChEBI CHEBI:29022
IUPAC Name N-benzyl-7H-purin-6-amine
SMILES C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 229.0°C to 233.0°C
Color White to Yellow
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Beilstein 26, IV, 3575
Formula Weight 225.25
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 6-Benzylaminopurine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468080 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number226410050,10328170,226410250,226410010
Lot NumberA0462420 Certificate TypeCertificate of Analysis Date13/05/2024 Catalog Number226410050,226410250,226410010
Lot NumberA0456590 Certificate TypeCertificate of Analysis Date26/09/2023 Catalog Number226410050,226410250,226410010
Lot NumberA0453191 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number226410050,226410250,226410010
Lot NumberA0450228 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number226410050,226410250,226410010
compliance-icons
Product Identifier
  • 6-Benzylaminopurine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Benzylaminopurine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.