missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-hexanol, 96%

Product Code. 11484817
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11484817

Brand: Thermo Scientific Alfa Aesar B21803.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4286-55-9 | C6H13BrO | 181.07 g/mol

A product for proteomics research. Also used as a reagent in the preparation of nitric oxide-donating analogues of Sulindac (S699215), a non-steroidal anti-inflammatory drug that has the ability to prevent colon cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A product for proteomics research. Also used as a reagent in the preparation of nitric oxide-donating analogues of Sulindac (S699215), a non-steroidal anti-inflammatory drug that has the ability to prevent colon cancer.

Solubility
Soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4286-55-9
Molecular Formula C6H13BrO
Molecular Weight (g/mol) 181.07
MDL Number MFCD00002983
InChI Key FCMCSZXRVWDVAW-UHFFFAOYSA-N
Synonym 6-bromo-1-hexanol, 6-bromohexanol, 1-hexanol, 6-bromo, 6-bromo-hexan-1-ol, hexamethylene bromohydrin, 6-bromo-hexanol, 6-bromo-l-hexanol, 1-bromo-6-hexanol, 1-bromohexan-6-ol, 6-hydroxyhexyl bromide
PubChem CID 77970
IUPAC Name 6-bromohexan-1-ol
SMILES OCCCCCCBr

Specifications

Density 1.384
Boiling Point 105°C to 106°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.482
Quantity 5 g
Solubility Information Soluble in chloroform and methanol.
Formula Weight 181.08
Percent Purity 97%
Chemical Name or Material 6-Bromo-1-hexanol

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis