missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1-hexene, 95%, Thermo Scientific Chemicals

Product Code. 11303436
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

6-Bromo-1-hexene was used in the preparation of epoxide, 6-bromo-1,2-epoxyhexane. It was also used in a simple, direct preparation of 4-alkenylbenzonitriles via anionic reduced forms of terephthalonitrile in liquid ammonia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-1-hexene was used in the preparation of epoxide, 6-bromo-1,2-epoxyhexane. It was also used in a simple, direct preparation of 4-alkenylbenzonitriles via anionic reduced forms of terephthalonitrile in liquid ammonia.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2695-47-8
Molecular Formula C6H11Br
Molecular Weight (g/mol) 163.058
MDL Number MFCD00000269
InChI Key RIMXEJYJXDBLIE-UHFFFAOYSA-N
Synonym 6-bromo-1-hexene, 5-hexenyl bromide, 1-hexene, 6-bromo, 1-bromo-5-hexene, 6-bromohexene, 6-bromo-hexene, 6-bromo-hex-1-ene, l-bromo-5-hexene, 6-bromo-1-hexen, 1-bromohex-5-ene
PubChem CID 75906
IUPAC Name 6-bromohex-1-ene
SMILES C=CCCCCBr

Specifications

Density 1.217
Boiling Point 147°C
Flash Point 54°C (129°F)
Odor Characteristic
Linear Formula Br(CH2)4CH=CH2
Refractive Index 1.4652
Quantity 5 g
UN Number UN1993
Beilstein 506203
Solubility Information Not miscible in water.
Formula Weight 163.06
Percent Purity 95%
Chemical Name or Material 6-Bromo-1-hexene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.