missing translation for 'onlineSavingsMsg'
Learn More

6-Bromochromone-2-carboxylic acid, 97%, Thermo Scientific™

86.23 € - 257.00 €

Chemical Identifiers

CAS 51484-06-1
Molecular Formula C10H5BrO4
Molecular Weight (g/mol) 269.05
MDL Number MFCD01242619
InChI Key QSBZDBNPXSVVHH-UHFFFAOYSA-N
Synonym 6-bromochromone-2-carboxylic acid, 6-bromo-4-oxo-4h-chromene-2-carboxylic acid, 6-bromo-4-oxo-4h-1-benzopyran-2-carboxylic acid, acmc-20alzt, 6-bromo-4-oxo-4h-chromene-2-carboxylicacid, 6-bromochromone-2-carboxylicacid, 6-brornochrornone-2-carboxylic acid
PubChem CID 688867
IUPAC Name 6-bromo-4-oxochromene-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)C2=CC(Br)=CC=C2O1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15424677
View Documents
Thermo Scientific Alfa Aesar
H32009.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15434677
View Documents
Thermo Scientific Alfa Aesar
H32009.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

51484-06-1
269.05
QSBZDBNPXSVVHH-UHFFFAOYSA-N
688867
OC(=O)C1=CC(=O)C2=CC(Br)=CC=C2O1
C10H5BrO4
MFCD01242619
6-bromochromone-2-carboxylic acid, 6-bromo-4-oxo-4h-chromene-2-carboxylic acid, 6-bromo-4-oxo-4h-1-benzopyran-2-carboxylic acid, acmc-20alzt, 6-bromo-4-oxo-4h-chromene-2-carboxylicacid, 6-bromochromone-2-carboxylicacid, 6-brornochrornone-2-carboxylic acid
6-bromo-4-oxochromene-2-carboxylic acid

Specifications

51484-06-1
C10H5BrO4
6-bromochromone-2-carboxylic acid, 6-bromo-4-oxo-4h-chromene-2-carboxylic acid, 6-bromo-4-oxo-4h-1-benzopyran-2-carboxylic acid, acmc-20alzt, 6-bromo-4-oxo-4h-chromene-2-carboxylicacid, 6-bromochromone-2-carboxylicacid, 6-brornochrornone-2-carboxylic acid
OC(=O)C1=CC(=O)C2=CC(Br)=CC=C2O1
269.05
269.05
6-Bromochromone-2-carboxylic acid
265°C
MFCD01242619
QSBZDBNPXSVVHH-UHFFFAOYSA-N
6-bromo-4-oxochromene-2-carboxylic acid
688867
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.