missing translation for 'onlineSavingsMsg'
Learn More
6-Bromoimidazo[1,2-a]pyridine, 98%, Thermo Scientific Chemicals Product Code.: 15467397

6-Bromoimidazo[1,2-a]pyridine, 98%, Thermo Scientific Chemicals

Product Code. 15467397
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15467397

Brand: Thermo Scientific Alfa Aesar H34453.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in organic syntheses and as pharmaceutical intermediates. The best results were obtained with lactams that could be introduced on the 6-bromoimidazo[1,2-a]pyridine. 1,2-dihydro-5-imidazo [1,2-a]pyridin-6-yl-6-methyl-2-oxo-3-pyridinecarbonitrile hydrochloride monohydrate 6 was synthesized in 5 steps from 6-bromoimidazo[1,2-a] pyridine using [2- 14 C] cyanoacetamide as the source of the radiolabel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic syntheses and as pharmaceutical intermediates. The best results were obtained with lactams that could be introduced on the 6-bromoimidazo[1,2-a]pyridine. 1,2-dihydro-5-imidazo [1,2-a]pyridin-6-yl-6-methyl-2-oxo-3-pyridinecarbonitrile hydrochloride monohydrate 6 was synthesized in 5 steps from 6-bromoimidazo[1,2-a] pyridine using [2- 14 C] cyanoacetamide as the source of the radiolabel.

Solubility
slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6188-23-4
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
MDL Number MFCD06496200
InChI Key FXPMFQUOGYGTAM-UHFFFAOYSA-N
Synonym 6-bromoimidazo 1,2-a pyridine, 6-bromoh-imidazo 1,2-a pyridine, 6-bromo-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine, 6-bromo, buttpark 154\50-43, pubchem14736, acmc-209mx4, 6-bromo-4-hydroimidazo 1,2-a pyridineShow MoreShow Less
PubChem CID 2795547
IUPAC Name 6-bromoimidazo[1,2-a]pyridine
SMILES C1=CC2=NC=CN2C=C1Br

Specifications

Melting Point 78°C to 80°C
Quantity 5 g
Solubility Information slightly soluble in water.
Formula Weight 197.03
Percent Purity 98%
Chemical Name or Material 6-Bromoimidazo[1,2-a]pyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10188398 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberH34453.03,H34453.06
Lot Number10188398 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH34453.03,H34453.06
Lot Number10165842 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH34453.03,H34453.06
Lot Number10172193 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH34453.03,H34453.06

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
6-Bromoimidazo[1,2-a]pyridine, 98%, Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado