Learn More
6-Chloro-2-fluoro-3-methylphenol, 97+%, Thermo Scientific™
45.00 € - 188.94 €
Chemical Identifiers
CAS | 261762-91-8 |
---|---|
Molecular Formula | C7H6ClFO |
Molecular Weight (g/mol) | 160.57 |
MDL Number | MFCD01631376 |
InChI Key | HCZRPIVKIPIGDM-UHFFFAOYSA-N |
PubChem CID | 2773684 |
IUPAC Name | 6-chloro-2-fluoro-3-methylphenol |
SMILES | CC1=CC=C(Cl)C(O)=C1F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11456326
|
Thermo Scientific Alfa Aesar
B23113.03 |
1 g | N/A | ||||||
11466326
|
Thermo Scientific Alfa Aesar
B23113.06 |
5 g |
188.94 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
261762-91-8 | |
160.57 | |
HCZRPIVKIPIGDM-UHFFFAOYSA-N | |
6-chloro-2-fluoro-3-methylphenol |
C7H6ClFO | |
MFCD01631376 | |
2773684 | |
CC1=CC=C(Cl)C(O)=C1F |
Specifications
261762-91-8 | |
C7H6ClFO | |
UN3261 | |
HCZRPIVKIPIGDM-UHFFFAOYSA-N | |
6-chloro-2-fluoro-3-methylphenol | |
2773684 | |
≥97% |
46°C to 49°C | |
MFCD01631376 | |
6-chloro-2-fluoro-m-cresol, acmc-20alzj, 6-chloro-2-fluoro-3-methyl phenol, 6-chloro-2-fluoro-3-methyl-phenol, phenol,6-chloro-2-fluoro-3-methyl, phenol, 6-chloro-2-fluoro-3-methyl | |
CC1=CC=C(Cl)C(O)=C1F | |
160.57 | |
160.58 | |
6-Chloro-2-fluoro-3-methylphenol |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures