missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2H-chromene-3-carbonyl chloride, 95%, Thermo Scientific™

53.10 € - 280.00 €

Chemical Identifiers

CAS 306935-54-6
Molecular Formula C10H6Cl2O2
Molecular Weight (g/mol) 229.056
MDL Number MFCD02091502
InChI Key HGFGWTNRUGRBNQ-UHFFFAOYSA-N
Synonym 6-chloro-2h-1-benzopyran-3-carbonyl chloride, 2h-1-benzopyran-3-carbonylchloride, 6-chloro, 6-chloro-2h-1-benzopyran-3-carbonylchloride, 6-chloranyl-2h-chromene-3-carbonyl chloride
PubChem CID 2735716
IUPAC Name 6-chloro-2H-chromene-3-carbonyl chloride
SMILES C1C(=CC2=C(O1)C=CC(=C2)Cl)C(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10140432
View Documents
Thermo Scientific Maybridge
RDR03724DA
1 g
53.10 €
1g
Please to purchase this item. Need a web account? Register with us today!
10304342
View Documents
Thermo Scientific Maybridge
RDR03724EA
10 g
280.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

306935-54-6
229.056
HGFGWTNRUGRBNQ-UHFFFAOYSA-N
2735716
C1C(=CC2=C(O1)C=CC(=C2)Cl)C(=O)Cl
C10H6Cl2O2
MFCD02091502
6-chloro-2h-1-benzopyran-3-carbonyl chloride, 2h-1-benzopyran-3-carbonylchloride, 6-chloro, 6-chloro-2h-1-benzopyran-3-carbonylchloride, 6-chloranyl-2h-chromene-3-carbonyl chloride
6-chloro-2H-chromene-3-carbonyl chloride

Specifications

306935-54-6
95%
MFCD02091502
HGFGWTNRUGRBNQ-UHFFFAOYSA-N
6-chloro-2H-chromene-3-carbonyl chloride
2735716
95%
95.0
C10H6Cl2O2
6-chloro-2h-1-benzopyran-3-carbonyl chloride, 2h-1-benzopyran-3-carbonylchloride, 6-chloro, 6-chloro-2h-1-benzopyran-3-carbonylchloride, 6-chloranyl-2h-chromene-3-carbonyl chloride
C1C(=CC2=C(O1)C=CC(=C2)Cl)C(=O)Cl
229.056
229.06
6-Chloro-2H-chromene-3-carbonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.