missing translation for 'onlineSavingsMsg'
Learn More
6-Chloro-3(2H)-pyridazinone, 98%, Thermo Scientific Chemicals Product Code.: 15478418

6-Chloro-3(2H)-pyridazinone, 98%, Thermo Scientific Chemicals

Product Code. 15478418
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15478418

Brand: Thermo Scientific Alfa Aesar H61837.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19064-67-6
Molecular Formula C4H3ClN2O
Molecular Weight (g/mol) 130.531
MDL Number MFCD00160460
InChI Key YICPBKWYZXFJNB-UHFFFAOYSA-N
Synonym 6-chloropyridazin-3-ol, 6-chloro-3-hydroxypyridazine, 6-chloro-2h-pyridazin-3-one, 6-chloropyridazin-3 2h-one, 3-chloro-6-hydroxypyridazine, 3 2h-pyridazinone, 6-chloro, 6-chloro-3 2h-pyridazinone, 3-chloro-6-pyridazone, 3-hydroxy-6-chloropyridazine, 6-chloro-3-pyridazinol
PubChem CID 252828
IUPAC Name 3-chloro-1H-pyridazin-6-one
SMILES C1=CC(=NNC1=O)Cl

Specifications

Quantity 5 g
Formula Weight 130.53
Percent Purity 98%
Chemical Name or Material 6-Chloro-3(2H)-pyridazinone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP15I027 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH61837.06
Lot NumberN07E007 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61837.06
Lot Number81202681 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61837.06
Lot NumberQ24F068 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61837.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Chloro-3(2H)-pyridazinone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.