missing translation for 'onlineSavingsMsg'
Learn More
6-Chloro-3-indolyl-beta-D-galactopyranoside, 98%, Thermo Scientific Chemicals Product Code.: 11494617

6-Chloro-3-indolyl-beta-D-galactopyranoside, 98%, Thermo Scientific Chemicals

Product Code. 11494617
0.1 g
Click to view available options
Quantity:
0.1 g
0.5 g
This item is not returnable. View return policy

Product Code. 11494617

Brand: Thermo Scientific Alfa Aesar B21131.77

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a pharmaceutical intermediate to reduce toxicity and as an enzyme substrate in diagnostic reagents. Salmon-gal is used in conjunction with IPTG for detection of β-galactosidase activity in bacterial colonies in a colometric assay to detect recombinants (white) from the non-recombinants (SALMON). In conjunction with X-glu, salmon-gal is useful for simultaneous detection of GUS and Lac activities on the same plate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a pharmaceutical intermediate to reduce toxicity and as an enzyme substrate in diagnostic reagents. Salmon-gal is used in conjunction with IPTG for detection of β-galactosidase activity in bacterial colonies in a colometric assay to detect recombinants (white) from the non-recombinants (SALMON). In conjunction with X-glu, salmon-gal is useful for simultaneous detection of GUS and Lac activities on the same plate.

Solubility
Insoluble in water. Soluble in DMSO and DMFO, methanol.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138182-21-5
Molecular Formula C14H16ClNO6
Molecular Weight (g/mol) 329.73
MDL Number MFCD00467206,MFCD01310928
InChI Key OQWBAXBVBGNSPW-MBJXGIAVSA-N
Synonym salmon-gal, rose-gal, 6-chloro-3-indolyl beta-d-galactopyranoside
PubChem CID 44630036
IUPAC Name (2S,3S,4S,5R,6S)-2-[(6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=CC=C23)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Melting Point 188°C to 190°C
Quantity 0.1 g
Sensitivity Moisture sensitive
Solubility Information Insoluble in water. Soluble in DMSO and DMFO,methanol.
Optical Rotation −47° (c=1 in Methanol)
Formula Weight 329.74
Percent Purity 98%
Chemical Name or Material 6-Chloro-3-indolyl-beta-D-galactopyranoside

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10216439 Certificate TypeCertificate of Analysis Date30/06/2023 Catalog NumberB21131.02,B21131.77,B21131
Lot Number10204290 Certificate TypeCertificate of Analysis Date30/06/2023 Catalog NumberB21131.02,B21131.77,B21131
Lot Number10195267 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21131.02,B21131.77
Lot Number10204290 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21131.02,B21131.77
Lot Number10216439 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21131.02,B21131.77

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Chloro-3-indolyl-beta-D-galactopyranoside, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.