missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorochroman-4-one, 99%, Thermo Scientific Chemicals

33.90 € - 566.00 €

Chemical Identifiers

CAS 37674-72-9
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.603
MDL Number MFCD00024070
InChI Key LLTDYHFVIVSQPJ-UHFFFAOYSA-N
Synonym 6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5
PubChem CID 95273
IUPAC Name 6-chloro-2,3-dihydrochromen-4-one
SMILES C1COC2=C(C1=O)C=C(C=C2)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11447187
View Documents
Thermo Scientific Alfa Aesar
A14975.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11457187
View Documents
Thermo Scientific Alfa Aesar
A14975.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11467187
View Documents
Thermo Scientific Alfa Aesar
A14975.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

6-Chlorochroman-4-one is used as chemical, organic intermediates for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Chlorochroman-4-one is used as chemical, organic intermediates for pharmaceuticals.

Solubility
Soluble in solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

37674-72-9
182.603
LLTDYHFVIVSQPJ-UHFFFAOYSA-N
95273
C1COC2=C(C1=O)C=C(C=C2)Cl
C9H7ClO2
MFCD00024070
6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5
6-chloro-2,3-dihydrochromen-4-one

Specifications

37674-72-9
C9H7ClO2
124218
Soluble in solvents.
C1COC2=C(C1=O)C=C(C=C2)Cl
182.603
182.61
6-Chlorochroman-4-one
101°C to 102°C
MFCD00024070
6-chlorochroman-4-one, 6-chloro-4-chromanone, 4h-1-benzopyran-4-one, 6-chloro-2,3-dihydro, 6-chloro chroman-4-one, 6-chloro-2,3-dihydro-4h-chromen-4-one, 6-chloro-2,3-dihydro-1-benzopyran-4-one, 6-chloro-3,4-dihydro-2h-1-benzopyran-4-one, pubchem17931, 6-chloro-chroman-4-one, acmc-1acq5
LLTDYHFVIVSQPJ-UHFFFAOYSA-N
6-chloro-2,3-dihydrochromen-4-one
95273
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 253-611-1

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.