missing translation for 'onlineSavingsMsg'
Learn More

6-Chlorohexanoyl chloride, 95%, Thermo Scientific Chemicals

Product Code. 11347566
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19347-73-0
Molecular Formula C6H10Cl2O
Molecular Weight (g/mol) 169.045
MDL Number MFCD00041436
InChI Key WZILXAPNPKMOSA-UHFFFAOYSA-N
Synonym hexanoyl chloride, 6-chloro, 6-chloro-caproyl chloride, acmc-1bt4p, 6-chloro-hexanoyl chloride, 6-chlorocaproic acid chloride, 6-chlorohexanoyl chloride
PubChem CID 88014
IUPAC Name 6-chlorohexanoyl chloride
SMILES C(CCC(=O)Cl)CCCl

Specifications

Boiling Point 90°C (7 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN3265
Beilstein 1852393
Formula Weight 169.05
Percent Purity 95%
Chemical Name or Material 6-Chlorohexanoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'