missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Chloroindole, 99%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 88°C to 91°C |
Color | Beige |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Quantity | 250 mg |
Solubility Information | Solubility in water: slightly soluble |
Formula Weight | 151.6 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'