missing translation for 'onlineSavingsMsg'
Learn More

[6-(Diethylamino)-3-pyridinyl]methanol, 97%, Thermo Scientific™

Product Code. 10289742
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10289742

Brand: Thermo Scientific Maybridge MO07117DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-68-9
Molecular Formula C10H16N2O
Molecular Weight (g/mol) 180.251
MDL Number MFCD06200887
InChI Key OMGIPGLZKJOJSG-UHFFFAOYSA-N
Synonym 6-diethylamino-3-pyridinyl methanol, 6-diethylamino pyridin-3-yl methanol, 6-diethylamino-3-pyridyl methan-1-ol
PubChem CID 2794793
IUPAC Name [6-(diethylamino)pyridin-3-yl]methanol
SMILES CCN(CC)C1=NC=C(C=C1)CO

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 180.25
Percent Purity 97%
Chemical Name or Material [6-(Diethylamino)-3-pyridinyl]methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.