missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-4H-1,3-benzodioxine-8-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10481664
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 321309-28-8
Molecular Formula C9H7FO4
Molecular Weight (g/mol) 198.149
MDL Number MFCD02681981
InChI Key HWBALMSPYAUMMB-UHFFFAOYSA-N
Synonym 6-fluoro-4h-benzo 1,3 dioxine-8-carboxylic acid, 6-fluoro-2,4-dihydro-1,3-benzodioxine-8-carboxylic acid, 6-fluoro-4h-benzo d 1,3 dioxine-8-carboxylic acid, 6-fluoro-4h-1,3-benzodioxin-8-carboxylic acid, 8-carboxy-6-fluoro-4h-1,3-benzodioxine, 6-fluoro-1,3-benzodioxene-8-carboxylic acid, 4h-1,3-benzodioxin-8-carboxylicacid, 6-fluoro, 6-fluoranyl-4h-1,3-benzodioxine-8-carboxylic acid, 6-fluoro-2h,4h-benzo e 1,3-dioxane-8-carboxylic acid, fbg
PubChem CID 2779901
IUPAC Name 6-fluoro-4H-1,3-benzodioxine-8-carboxylic acid
SMILES C1C2=C(C(=CC(=C2)F)C(=O)O)OCO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 198.15
Percent Purity 97%
Chemical Name or Material 6-Fluoro-4H-1, 3-benzodioxine-8-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'