missing translation for 'onlineSavingsMsg'
Learn More
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline, 97%, Thermo Scientific Chemicals Product Code.: 15451759

6-Hydroxy-1,2,3,4-tetrahydroisoquinoline, 97%, Thermo Scientific Chemicals

Product Code. 15451759
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15451759

Brand: Thermo Scientific Alfa Aesar H64512.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14446-24-3
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD01717047
InChI Key SCMZIFSYPJICCV-UHFFFAOYSA-N
Synonym 6-hydroxy-1,2,3,4-tetrahydroisoquinoline, 1,2,3,4-tetrahydro-isoquinolin-6-ol, 1,2,3,4-tetrahydro-6-isoquinolinol, 6-isoquinolinol, 1,2,3,4-tetrahydro, pubchem13941, 6-hydroxytetrahydroisochinolin, ksc376k4j, 6-hydroxy-1,2,3,4-tetrahydro-isoquinolineShow MoreShow Less
PubChem CID 26698
IUPAC Name 1,2,3,4-tetrahydroisoquinolin-6-ol
SMILES C1CNCC2=C1C=C(C=C2)O

Specifications

Melting Point 192°C to 193°C
Quantity 250 mg
Beilstein 127578
Formula Weight 149.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Hydroxy-1,2,3,4-tetrahydroisoquinoline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81400914 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH64512.MD,H64512.03
compliance-icons
Product Identifier
  • 6-Hydroxy-1,2,3,4-tetrahydroisoquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 6-Hydroxy-1,2,3,4-tetrahydroisoquinoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Hydroxy-1,2,3,4-tetrahydroisoquinoline, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.