missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2,3-dihydrobenzo[b]furan-3-one, 97%, Thermo Scientific™

Product Code. 10273372
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6272-26-0
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00068174
InChI Key GBDMODVZBPFQKI-UHFFFAOYSA-N
Synonym 6-hydroxybenzofuran-3 2h-one, 6-hydroxy-2h-benzofuran-3-one, 6-hydroxy-2,3-dihydrobenzo b furan-3-one, 6-hydroxy-1-benzofuran-3 2h-one, 6-hydroxybenzofuran-3-one, 3 2h-benzofuranone, 6-hydroxy, 6-hydroxy-3-coumaranone, 6-hydroxy-2,3-dihydro-1-benzofuran-3-one, 6-hydroxy-3 2h-benzofuranone, 6-hydroxy-2h-1-benzofuran-3-one
PubChem CID 235717
IUPAC Name 6-hydroxy-1-benzofuran-3-one
SMILES OC1=CC=C2C(=O)COC2=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 150.13
Percent Purity 97%
Chemical Name or Material 6-Hydroxy-2, 3-dihydrobenzo[b]furan-3-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'