missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-4-methylcoumarin, 99%, Thermo Scientific™

Product Code. 11347176
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2373-31-1
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00016967
InChI Key IRUHWRSITUYICV-UHFFFAOYSA-N
PubChem CID 75409
IUPAC Name 6-hydroxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=C1C=C(C=C2)O

Specifications

Melting Point 246°C to 250°C
Quantity 1 g
Beilstein 144873
Synonym 6-hydroxy-4-methylcoumarin, 6-hydroxy-4-methyl-2h-chromen-2-one, coumarin, 6-hydroxy-4-methyl, 6-hydroxy-4-methyl-2-benzopyrone, 6-hydroxy-4-methyl-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy-4-methyl, chembl12626, 2h-1-benzopyran-2-one,6-hydroxy-4-methyl, acmc-1cuho, 4-methyl-6-hydroxycoumarin
Formula Weight 176.17
Percent Purity 99%
Chemical Name or Material 6-Hydroxy-4-methylcoumarin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'