missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxycoumarin, 98%, Thermo Scientific Chemicals

25.90 € - 191.36 €

Chemical Identifiers

CAS 6093-68-1
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.15
InChI Key CJIJXIFQYOPWTF-UHFFFAOYSA-N
Synonym 6-hydroxycoumarin, 6-hydroxy-2h-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy, 6-hydroxycoumaran, 2h-1-benzopyran-2-one,6-hydroxy, 6-hydroxbenzopyran-2-one, 6-hydroxy-2h-chromen-2-on, 6-hydroxycoumarine, 6-hydroxy coumarin, 6-hydroxy-2h-1-benzopyran-2-one
PubChem CID 99477
IUPAC Name 6-hydroxychromen-2-one
SMILES C1=CC2=C(C=CC(=O)O2)C=C1O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15590624
View Documents
Thermo Scientific Acros
459410010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15500634
View Documents
Thermo Scientific Acros
459410050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15510634
View Documents
Thermo Scientific Acros
459410250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6093-68-1
162.15
6-hydroxycoumarin, 6-hydroxy-2h-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy, 6-hydroxycoumaran, 2h-1-benzopyran-2-one,6-hydroxy, 6-hydroxbenzopyran-2-one, 6-hydroxy-2h-chromen-2-on, 6-hydroxycoumarine, 6-hydroxy coumarin, 6-hydroxy-2h-1-benzopyran-2-one
6-hydroxychromen-2-one
C9H6O3
CJIJXIFQYOPWTF-UHFFFAOYSA-N
99477
C1=CC2=C(C=CC(=O)O2)C=C1O

Specifications

6093-68-1
Authentic
Glass Bottle
6-hydroxycoumarin, 6-hydroxy-2h-chromen-2-one, 2h-1-benzopyran-2-one, 6-hydroxy, 6-hydroxycoumaran, 2h-1-benzopyran-2-one,6-hydroxy, 6-hydroxbenzopyran-2-one, 6-hydroxy-2h-chromen-2-on, 6-hydroxycoumarine, 6-hydroxy coumarin, 6-hydroxy-2h-1-benzopyran-2-one
CJIJXIFQYOPWTF-UHFFFAOYSA-N
6-hydroxychromen-2-one
99477
98%
249.0°C to 253.0°C
97.5% min. (HPLC)
C9H6O3
Solubility in water: insoluble.
C1=CC2=C(C=CC(=O)O2)C=C1O
162.15
162.15
6-Hydroxycoumarin
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

Product Identifier
  • 6-Hydroxycoumarin

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only