missing translation for 'onlineSavingsMsg'
Learn More
6-Hydroxydopamine hydrobromide, Tocris Bioscience™ Product Code.: 18044622

6-Hydroxydopamine hydrobromide, Tocris Bioscience™

Product Code. 18044622
50 mg
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy

Product Code. 18044622

Brand: Tocris Bioscience 2547/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective catecholaminergic neurotoxin

Selective catecholaminergic neurotoxin. Depletes brain catecholamine levels via uptake and accumulation by a transport mechanism specific to these neurons. Causes almost complete destruction of nigral dopaminergic neurons and their striatal terminals when injected into the substantia nigra of rats, producing an animal model of Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

CAS 636-00-0
Molecular Formula C8H12BrNO3
Quantity 50 mg
Synonym 6-hydroxydopamine hydrobromide, oxidopamine hydrobromide, 6-hydroxydopamine hydrogen bromide, unii-twc1d4w0wb, 6-hydroxydopamine hbr, 5-2-aminoethyl benzene-1,2,4-triol hydrobromide, twc1d4w0wb, hydroxydopamine hydrobromide, 6-oh-da
InChI Key MLACDGUOKDOLGC-UHFFFAOYSA-N
SMILES C1=C(C(=CC(=C1O)O)O)CCN.Br
IUPAC Name 5-(2-aminoethyl)benzene-1,2,4-triol;hydrobromide
Molecular Weight (g/mol) 250.092
PubChem CID 176170
Formula Weight 250.09
Percent Purity >98%
Chemical Name or Material 6-Hydroxydopamine hydrobromide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Hydroxydopamine hydrobromide, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.