missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1,2,3,4-tetrahydroquinoline, 97%, Thermo Scientific Chemicals

Product Code. 15469428
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-15-0
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00090494
InChI Key FRXSZNDVFUDTIR-UHFFFAOYSA-N
Synonym thallin, thalline, 6-methoxy-1,2,3,4-tetrahydro-quinoline, 6-methoxytetrahydroquinoline, quinoline, 1,2,3,4-tetrahydro-6-methoxy, 1,2,3,4-tetrahydro-6-methoxyquinoline, methyl 1,2,3,4-tetrahydro-6-quinolyl ether, quinoline l,2,3,4-tetrahydro-6-methoxy, wln: t66 bmt & j ho1, 1,3,4-tetrahydro-6-methoxyquinoline
PubChem CID 67113
IUPAC Name 6-methoxy-1,2,3,4-tetrahydroquinoline
SMILES COC1=CC2=C(C=C1)NCCC2

Specifications

Melting Point 34°C to 37°C
Quantity 5 g
Formula Weight 163.22
Percent Purity 97%
Chemical Name or Material 6-Methoxy-1,2,3,4-tetrahydroquinoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.