missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-1,2,3,4-tetrahydroquinoline, 97%, Thermo Scientific Chemicals

Product Code. 15479428
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2,3,4-Tetrahydro-6-methoxyquinoline is used as chemical reagents, organic intermediates, fine chemicals, pharmaceutical research and development.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-15-0
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00090494
InChI Key FRXSZNDVFUDTIR-UHFFFAOYSA-N
Synonym thallin, thalline, 6-methoxy-1,2,3,4-tetrahydro-quinoline, 6-methoxytetrahydroquinoline, quinoline, 1,2,3,4-tetrahydro-6-methoxy, 1,2,3,4-tetrahydro-6-methoxyquinoline, methyl 1,2,3,4-tetrahydro-6-quinolyl ether, quinoline l,2,3,4-tetrahydro-6-methoxy, wln: t66 bmt & j ho1, 1,3,4-tetrahydro-6-methoxyquinoline
PubChem CID 67113
IUPAC Name 6-methoxy-1,2,3,4-tetrahydroquinoline
SMILES COC1=CC2=C(C=C1)NCCC2

Specifications

Melting Point 34°C to 37°C
Quantity 250 mg
Formula Weight 163.22
Percent Purity 97%
Chemical Name or Material 6-Methoxy-1,2,3,4-tetrahydroquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'