missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxynaphthalene-2-boronic acid, 95%, Thermo Scientific Chemicals

27.84 € - 80.30 €

Chemical Identifiers

CAS 156641-98-4
Molecular Formula C11H11BO3
Molecular Weight (g/mol) 202.02
MDL Number MFCD03093087
InChI Key GZFAVYWCPSMLCM-UHFFFAOYSA-N
Synonym 6-methoxy-2-naphthaleneboronic acid, 6-methoxynaphthalen-2-yl boronic acid, 6-methoxy-2-naphthylboronic acid, 6-methoxynaphthalene-2-boronic acid, 6-methoxy-2-naphthyl boronic acid, 2-methoxynaphthalene-6-boronic acid, boronic acid, 6-methoxy-2-naphthalenyl, 6-methoxy-2-naphthalenyl boronic acid, acmc-209deh, ksc489k9r
PubChem CID 4641692
IUPAC Name (6-methoxynaphthalen-2-yl)boronic acid
SMILES COC1=CC2=CC=C(C=C2C=C1)B(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11319605
View Documents
Thermo Scientific Alfa Aesar
L19060.03
1 g
27.84 €
1g
Please to purchase this item. Need a web account? Register with us today!
11329605
View Documents
Thermo Scientific Alfa Aesar
L19060.06
5 g
80.30 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

156641-98-4
202.02
GZFAVYWCPSMLCM-UHFFFAOYSA-N
4641692
COC1=CC2=CC=C(C=C2C=C1)B(O)O
C11H11BO3
MFCD03093087
6-methoxy-2-naphthaleneboronic acid, 6-methoxynaphthalen-2-yl boronic acid, 6-methoxy-2-naphthylboronic acid, 6-methoxynaphthalene-2-boronic acid, 6-methoxy-2-naphthyl boronic acid, 2-methoxynaphthalene-6-boronic acid, boronic acid, 6-methoxy-2-naphthalenyl, 6-methoxy-2-naphthalenyl boronic acid, acmc-209deh, ksc489k9r
(6-methoxynaphthalen-2-yl)boronic acid

Specifications

156641-98-4
C11H11BO3
7636998
GZFAVYWCPSMLCM-UHFFFAOYSA-N
(6-methoxynaphthalen-2-yl)boronic acid
4641692
95%
∽245°C (decomposition)
MFCD03093087
6-methoxy-2-naphthaleneboronic acid, 6-methoxynaphthalen-2-yl boronic acid, 6-methoxy-2-naphthylboronic acid, 6-methoxynaphthalene-2-boronic acid, 6-methoxy-2-naphthyl boronic acid, 2-methoxynaphthalene-6-boronic acid, boronic acid, 6-methoxy-2-naphthalenyl, 6-methoxy-2-naphthalenyl boronic acid, acmc-209deh, ksc489k9r
COC1=CC2=CC=C(C=C2C=C1)B(O)O
202.02
202.02
6-Methoxynaphthalene-2-boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.