Learn More
6-Methoxypyridine-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals
141.00 € - 413.00 €
Chemical Identifiers
CAS | 312300-42-8 |
---|---|
Molecular Formula | C6H6ClNO3S |
Molecular Weight (g/mol) | 207.63 |
MDL Number | MFCD06739108 |
InChI Key | OMLIVJOTRYWHNY-UHFFFAOYSA-N |
Synonym | 6-methoxy-pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-6-methoxypyridine, 6-methoxypyridin-3-ylsulfonyl chloride, 2-methoxypyridine-5-sulfonylchloride, 6-methoxy-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride, 6-methoxy, 6-methoxypyridine-3-sulphonyl chloride, 5-chlorosulfonyl-2-methoxypyridine, 6-methoxypyridine-3-yl sulfonyl chloride |
PubChem CID | 22344709 |
IUPAC Name | 6-methoxypyridine-3-sulfonyl chloride |
SMILES | COC1=NC=C(C=C1)S(Cl)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10564028
|
Thermo Scientific Acros
429410010 |
1 g | Glass bottle |
141.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10155857
|
Thermo Scientific Acros
429410050 |
5 g | Glass bottle |
413.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
312300-42-8 | |
207.63 | |
OMLIVJOTRYWHNY-UHFFFAOYSA-N | |
22344709 | |
COC1=NC=C(C=C1)S(Cl)(=O)=O |
C6H6ClNO3S | |
MFCD06739108 | |
6-methoxy-pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-6-methoxypyridine, 6-methoxypyridin-3-ylsulfonyl chloride, 2-methoxypyridine-5-sulfonylchloride, 6-methoxy-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride, 6-methoxy, 6-methoxypyridine-3-sulphonyl chloride, 5-chlorosulfonyl-2-methoxypyridine, 6-methoxypyridine-3-yl sulfonyl chloride | |
6-methoxypyridine-3-sulfonyl chloride |
Specifications
312300-42-8 | |
100.0 | |
White | |
96% min. (GC) | |
MFCD06739108 | |
OMLIVJOTRYWHNY-UHFFFAOYSA-N | |
6-methoxypyridine-3-sulfonyl chloride | |
22344709 | |
97% | |
6-Methoxypyridine-3-sulfonyl chloride |
96.0 | |
42.0°C to 45.0°C | |
Authentic | |
C6H6ClNO3S | |
6-methoxy-pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-6-methoxypyridine, 6-methoxypyridin-3-ylsulfonyl chloride, 2-methoxypyridine-5-sulfonylchloride, 6-methoxy-3-pyridinesulfonyl chloride, 3-pyridinesulfonylchloride, 6-methoxy, 6-methoxypyridine-3-sulphonyl chloride, 5-chlorosulfonyl-2-methoxypyridine, 6-methoxypyridine-3-yl sulfonyl chloride | |
COC1=NC=C(C=C1)S(Cl)(=O)=O | |
207.63 | |
207.64 | |
Crystalline Low Melting Solid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
- 6-Methoxypyridine-3-sulfonyl chloride, 97%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.