Learn More
6-Methyl-1,2,3,4-tetrahydroquinoline, 98%, Thermo Scientific Chemicals
154.00 € - 608.00 €
Chemical Identifiers
CAS | 91-61-2 |
---|---|
Molecular Formula | C10H13N |
Molecular Weight (g/mol) | 147.22 |
MDL Number | MFCD00023887 |
InChI Key | XOKMRXSMOHCNIX-UHFFFAOYSA-N |
Synonym | civettal, 1,2,3,4-tetrahydro-6-methylquinoline, quinoline, 1,2,3,4-tetrahydro-6-methyl, quinoline, tetrahydro-6-methyl, p-methyltetrahydroquinoline, acmc-209rck, dsstox_cid_27444, dsstox_rid_82351, dsstox_gsid_47444, xokmrxsmohcnix-uhfffaoysa |
PubChem CID | 66678 |
IUPAC Name | 6-methyl-1,2,3,4-tetrahydroquinoline |
SMILES | CC1=CC=C2NCCCC2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11442298
|
Thermo Scientific Alfa Aesar
A15245.09 |
10 g |
154.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11452298
|
Thermo Scientific Alfa Aesar
A15245.18 |
50 g |
608.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-Methyl-1,2,3,4-tetrahydroquinoline is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
91-61-2 | |
147.22 | |
XOKMRXSMOHCNIX-UHFFFAOYSA-N | |
66678 | |
CC1=CC=C2NCCCC2=C1 |
C10H13N | |
MFCD00023887 | |
civettal, 1,2,3,4-tetrahydro-6-methylquinoline, quinoline, 1,2,3,4-tetrahydro-6-methyl, quinoline, tetrahydro-6-methyl, p-methyltetrahydroquinoline, acmc-209rck, dsstox_cid_27444, dsstox_rid_82351, dsstox_gsid_47444, xokmrxsmohcnix-uhfffaoysa | |
6-methyl-1,2,3,4-tetrahydroquinoline |
Specifications
91-61-2 | |
262°C to 263°C (712 mmHg) | |
MFCD00023887 | |
civettal, 1,2,3,4-tetrahydro-6-methylquinoline, quinoline, 1,2,3,4-tetrahydro-6-methyl, quinoline, tetrahydro-6-methyl, p-methyltetrahydroquinoline, acmc-209rck, dsstox_cid_27444, dsstox_rid_82351, dsstox_gsid_47444, xokmrxsmohcnix-uhfffaoysa | |
XOKMRXSMOHCNIX-UHFFFAOYSA-N | |
6-methyl-1,2,3,4-tetrahydroquinoline | |
66678 | |
98% |
33°C to 37°C | |
C10H13N | |
122515 | |
Insoluble in water. | |
CC1=CC=C2NCCCC2=C1 | |
147.22 | |
147.22 | |
6-Methyl-1,2,3,4-tetrahydroquinoline |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
missing translation for 'einecsNumber' : 202-083-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only