missing translation for 'onlineSavingsMsg'
Learn More
6-methyl-2,3-dihydropyridazin-3-one hydrate, Thermo Scientific™ Product Code.: 10537893

6-methyl-2,3-dihydropyridazin-3-one hydrate, Thermo Scientific™

Product Code. 10537893
1 g
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 10537893

Brand: Thermo Scientific Maybridge BTB06840DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7143-82-0
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00039720
InChI Key QZWIXLPWMGHDDD-UHFFFAOYSA-N
Synonym 6-methylpyridazin-3-ol hydrate, 6-methylpyridazin-3 2h-one hydrate, 6-methyl-2,3-dihydropyridazin-3-one hydrate, 6-methyl-2h-pyridazin-3-one hydrate, 6-methylpyridazin-3-one hydrate, 3-methyl-1h-pyridazin-6-one hydrate, 6-methylpyridazin-3 2h ;-one hydrate, 6-methyl-2-hydropyridazin-3-one, hydrate, 6-methyl-2,3-dihydropyridazine-3-one hydrate, 6-methylpyridazin-3 2h-one-water 1/1Show MoreShow Less
PubChem CID 2774632
SMILES CC1=NNC(=O)C=C1

Specifications

Assay Percent Range 90%
Quantity 1 g
IUPAC Name 6-methyl-2,3-dihydropyridazin-3-one
Formula Weight 128.14
Percent Purity 90%
Chemical Name or Material 6-Methyl-2, 3-dihydropyridazin-3-one hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-methyl-2,3-dihydropyridazin-3-one hydrate, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.