Learn More
6-Methylsalicylic acid, 98%, Thermo Scientific Chemicals
45.20 € - 432.00 €
Chemical Identifiers
CAS | 567-61-3 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.15 |
MDL Number | MFCD01194284 |
InChI Key | HCJMNOSIAGSZBM-UHFFFAOYSA-N |
Synonym | 6-methylsalicylic acid, 6-hydroxy-o-toluic acid, 2,6-cresotic acid, 6-msa, benzoic acid, 2-hydroxy-6-methyl, 6-ms, methylsalicylic acid, 2-hydroxy-6-methylbenzoicacid, unii-l5352fe23y, 2-carboxy-3-methylphenol |
PubChem CID | 11279 |
ChEBI | CHEBI:17637 |
IUPAC Name | 2-hydroxy-6-methylbenzoic acid |
SMILES | CC1=CC=CC(O)=C1C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15302042
|
Thermo Scientific Acros
453060010 |
1 g |
45.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15312042
|
Thermo Scientific Acros
453060050 |
5 g |
139.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15322042
|
Thermo Scientific Acros
453060250 |
25 g |
432.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
567-61-3 | |
152.15 | |
HCJMNOSIAGSZBM-UHFFFAOYSA-N | |
11279 | |
2-hydroxy-6-methylbenzoic acid |
C8H8O3 | |
MFCD01194284 | |
6-methylsalicylic acid, 6-hydroxy-o-toluic acid, 2,6-cresotic acid, 6-msa, benzoic acid, 2-hydroxy-6-methyl, 6-ms, methylsalicylic acid, 2-hydroxy-6-methylbenzoicacid, unii-l5352fe23y, 2-carboxy-3-methylphenol | |
CHEBI:17637 | |
CC1=CC=CC(O)=C1C(O)=O |
Specifications
567-61-3 | |
Authentic | |
Glass Bottle | |
MFCD01194284 | |
HCJMNOSIAGSZBM-UHFFFAOYSA-N | |
2-hydroxy-6-methylbenzoic acid | |
11279 | |
152.15 | |
6-Methylsalicylic acid |
170.0°C to 175.0°C | |
97.5% min. (HPLC) | |
C8H8O3 | |
6-methylsalicylic acid, 6-hydroxy-o-toluic acid, 2,6-cresotic acid, 6-msa, benzoic acid, 2-hydroxy-6-methyl, 6-ms, methylsalicylic acid, 2-hydroxy-6-methylbenzoicacid, unii-l5352fe23y, 2-carboxy-3-methylphenol | |
CC1=CC=CC(O)=C1C(O)=O | |
152.15 | |
CHEBI:17637 | |
98% |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
- 6-Methylsalicylic acid, 98%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only