Learn More
BTX™ 6-needle Array Electrodes for BTX™ AgilePulse™ In Vivo
6-needle Array Electrodes for BTX™ AgilePulse™ In Vivo are miniature needles designed to provide highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse In Vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.
432.00 € - 585.00 €
Specifications
For Use With (Application) | Gene and Vaccine Delivery |
---|---|
For Use With (Equipment) | AgilePulse™ in Vivo Gene Delivery Electroporation System |
Material | Stainless Steel |
Product Line | BTX Electroporation |
Sterility | Sterile |
Product Code | Brand | Size (Metric) Gap | Length (Metric) | Tip Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) Gap | Length (Metric) | Tip Type | Price | Quantity & Availability | |||
15407380
|
BTX™
470050 |
4 mm | 2 mm | Fine Point |
432.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15417380
|
BTX™
470060 |
6 mm | 2 mm | Fine Point |
432.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15427380
|
BTX™
470070 |
6 mm | 10 mm | Fine Point |
432.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15447380
|
BTX™
470086 |
6 mm | 16 mm | Trocar Point |
585.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15437380
|
BTX™
470080 |
6 mm | 25 mm | Trocar Point |
432.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.
The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.
- Uniform, reliable electric fields
- Miniature needles minimize tissue trauma
- Disposable and sterile
- Medical-grade plastic and surgical steel construction
- Safety-assured design
- Multiple configurations, sizes to choose from
Specifications
Gene and Vaccine Delivery | |
Stainless Steel | |
Sterile | |
Square Wave |
AgilePulse™ in Vivo Gene Delivery Electroporation System | |
BTX Electroporation | |
Electrode Connector Cable |
Intended for Research use only. Not for use in diagnostic or clinical procedures.