missing translation for 'onlineSavingsMsg'
Learn More

6-Nitrocoumarin, 98+%, Thermo Scientific Chemicals

Product Code. 11492517
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2725-81-7
Molecular Formula C9H5NO4
Molecular Weight (g/mol) 191.142
MDL Number MFCD00016973
InChI Key RMERXEXZXIVNBF-UHFFFAOYSA-N
PubChem CID 75944
IUPAC Name 6-nitrochromen-2-one
SMILES C1=CC2=C(C=CC(=O)O2)C=C1[N+](=O)[O-]

Specifications

Melting Point 186°C to 190°C
Odor Odorless
Quantity 5 g
Beilstein 166079
Synonym 6-nitrocoumarin, 6-nitro-2h-chromen-2-one, 6-nitro-2-benzopyrone, 2h-1-benzopyran-2-one, 6-nitro, acmc-20abd6, maybridge1_002226, amtgc084, 6-nitro-2h-chromen-2-one #, 2h-1-benzopyran-2-one,6-nitro, 6-hydroxy oxido amino-2h-chromen-2-one
Formula Weight 191.14
Percent Purity ≥98%
Chemical Name or Material 6-Nitrocoumarin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.