missing translation for 'onlineSavingsMsg'
Learn More

6-Nitroindoline, 98%, Thermo Scientific™

33.60 € - 124.00 €

Chemical Identifiers

CAS 19727-83-4
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.16
MDL Number MFCD00005710
InChI Key LTNYDSMDSLOMSM-UHFFFAOYSA-N
Synonym 6-nitroindoline, indoline, 6-nitro, 1h-indole, 2,3-dihydro-6-nitro, 6-nitro-indoline, 2,3-dihydro-6-nitro-1h-indole, ccris 3302, zlchem 357, pubchem7443
PubChem CID 29757
IUPAC Name 6-nitro-2,3-dihydro-1H-indole
SMILES [O-][N+](=O)C1=CC=C2CCNC2=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10255871
View Documents
Thermo Scientific Acros
128710010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10768942
View Documents
Thermo Scientific Acros
128710050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

19727-83-4
164.16
LTNYDSMDSLOMSM-UHFFFAOYSA-N
29757
[O-][N+](=O)C1=CC=C2CCNC2=C1
C8H8N2O2
MFCD00005710
6-nitroindoline, indoline, 6-nitro, 1h-indole, 2,3-dihydro-6-nitro, 6-nitro-indoline, 2,3-dihydro-6-nitro-1h-indole, ccris 3302, zlchem 357, pubchem7443
6-nitro-2,3-dihydro-1H-indole

Specifications

19727-83-4
100.0
Orange or Red
97.5% min. (GC)
MFCD00005710
Solubility in water: negligible. Other solubilities: soluble in acetone,ethyl acetate and methanol
[O-][N+](=O)C1=CC=C2CCNC2=C1
164.16
164.16
Crystalline Powder or Crystals
97.5
66°C to 70°C
Authentic
C8H8N2O2
6-nitroindoline, indoline, 6-nitro, 1h-indole, 2,3-dihydro-6-nitro, 6-nitro-indoline, 2,3-dihydro-6-nitro-1h-indole, ccris 3302, zlchem 357, pubchem7443
LTNYDSMDSLOMSM-UHFFFAOYSA-N
6-nitro-2,3-dihydro-1H-indole
29757
98%
6-Nitroindoline, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 243-257-6

missing translation for 'rtecsNumber' : NM1950000

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.