missing translation for 'onlineSavingsMsg'
Learn More
(6-Phenyl-3-pyridinyl)methanol, 97%, Thermo Scientific™ Product Code.: 10755915

(6-Phenyl-3-pyridinyl)methanol, 97%, Thermo Scientific™

Product Code. 10755915
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 10755915

Brand: Thermo Scientific Maybridge CC21109DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 4634-09-7
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.23
MDL Number MFCD03086132
InChI Key RXXIVABOKBNIEI-UHFFFAOYSA-N
Synonym 6-phenylpyridin-3-yl methanol, 6-phenyl-3-pyridinyl methanol, 3-pyridinemethanol, 6-phenyl, 6-phenyl-3-pyridylmethanol, 3-pyridinemethanol,6-phenyl, 6-phenyl-3-pyridyl-methanol, 2-phenyl-5-hydroxymethylpyridine, rxxivabokbniei-uhfffaoysa-n, 6-phenyl-3-pyridyl methan-1-ol
PubChem CID 22254216

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 185.23
Percent Purity 97%
Chemical Name or Material (6-Phenyl-3-pyridinyl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(6-Phenyl-3-pyridinyl)methanol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.