Learn More
SCHOTT AG Lighting and Imaging™ 6-Point Ring Light
Provides optimal lighting for contrast while reducing shadows. Mounts directly to objective by means of thumb screw. For use with Cold light source KL300LED
Brand:  SCHOTT AG Lighting and Imaging™ 155102
Product Code. 10467542
  • 251.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 6-point Ringlight with eco-friendly Puravis fiber optics
  • Fiber bundle protected with flexible metal PVC sheathing.
  • Housing made of rugged, black anodized aluminum.
  • Cost effective in comparison to annular ringlights
  • Illumination with reduced shadows, but still sufficient shadows for improved contrasts
Specifications
Specifications
Fiber Optic Ringlight
Stereo Microscopy illumination
23 mm
Eco-friendly Puravis Glass Optical Fibers
66 mm ID
Cold light source KL300LED
1000 mm