missing translation for 'onlineSavingsMsg'
Learn More

6-Quinolinylmethanol, 97%, Thermo Scientific™

Product Code. 10357192
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 100516-88-9
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD03789621
InChI Key YQEJIIUSNDZIGO-UHFFFAOYSA-N
Synonym 6-quinolinylmethanol, 6-quinolinemethanol, 6-hydroxymethylquinoline, quinolin-6-yl methanol, 6-quinolylmethanol, pubchem12427, quinolin-6-yl-methanol, 6-hydroxymethyl quinoline, 6-hydroxymethyl-quinoline
PubChem CID 1514385
IUPAC Name quinolin-6-ylmethanol
SMILES C1=CC2=C(C=CC(=C2)CO)N=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 6-Quinolinylmethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'