missing translation for 'onlineSavingsMsg'
Learn More
6-Thioguanine, 98%, Thermo Scientific Chemicals Product Code.: 11499177

6-Thioguanine, 98%, Thermo Scientific Chemicals

Product Code. 11499177
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11499177

Brand: Thermo Scientific Alfa Aesar B21280.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

6-Thioguanine acts as an antineoplastic and purine antimetabolite. It is also useful as an inhibition of stimulated expression of TNF-related apoptosis-inducing ligand (TRAIL) protein. It is involved in the treatment of acute leukemias and Psoriasis. Further, it is used for the treatment of ulcerative colitis and autoimmune diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dilute solutions of alkali hydroxides and hot alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154-42-7
Molecular Formula C5H5N5S
Molecular Weight (g/mol) 167.19
MDL Number MFCD00233553
InChI Key WYWHKKSPHMUBEB-UHFFFAOYSA-N
Synonym 6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiolShow MoreShow Less
PubChem CID 2723601
ChEBI CHEBI:9555
SMILES NC1=NC(=S)C2=C(N1)N=CN2

Specifications

Melting Point >360°C
Assay Percent Range 0.98
Quantity 1 g
UN Number UN2811
Beilstein 157765
Merck Index 14,9337
Solubility Information Soluble in dilute solutions of alkali hydroxides and hot alcohol. Insoluble in water.
IUPAC Name 2-amino-3,7-dihydropurine-6-thione
Formula Weight 167.19
Percent Purity 98%
Chemical Name or Material 6-Thioguanine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252136 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog NumberB21280.06,B21280.03
Lot Number10249723 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog NumberB21280.06,B21280.03
Lot Number10247572 Certificate TypeCertificate of Analysis Date15/08/2023 Catalog NumberB21280.06,B21280.03
Lot NumberN21J018 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberB21280.03
Lot Number10244982 Certificate TypeCertificate of Analysis Date09/01/2023 Catalog NumberB21280.06,B21280.03
compliance-icons compliance-icons
Product Identifier
  • 6-Thioguanine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : Thioguanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Thioguanine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.