missing translation for 'onlineSavingsMsg'
Learn More
Learn More
600 mV Redox Oxidation/Reduction (ORP) Standard at 25°C, Reagecon
Acts as a control or reference material, rather than as a calibration standard because redox oxidation/reduction (ORP) is an absolute measurement and the sensing electrode does not require calibration.
Brand: Reagecon RS600
This item is not returnable.
View return policy
Description
- Redox Reference Standard, Redox Reference Material.
- Acts as a control or reference material rather than a calibration standard because the redox reaction is an absolute measurement and the sensing electrode does not require calibration.
- Controls the functionality of the sensing and reference electrode, the analysts technique, environmental conditions and the operation of the measuring meter.
- This 600 mV Redox (ORP) Standard is accurate to ± 5mV, has guaranteed stability throughout its entire shelf life, even after opening the bottle.
- The Value of 600 mV quoted refers to the potentials of the platinum electrode v Ag/AgCl reference electrode (3M KCl).
Specifications
Accuracy ± 5mV at 25°C | |
HDPE Bottle GL45 | |
600 mV (± 5mV) |
600 mV (± 5mV) | |
500 mL | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- redox standard 600mv @25
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, irontrichloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction