Learn More
YSI™ 60510 Pro Series ISE Cable
The 60510 is a 1-port cable for use with the Professional Plus instrument. The port requires a ISE probe.
368.00 € - 569.00 €
Product Code | Brand | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (English) | Length (Metric) | Price | Quantity & Availability | ||||
12963487
|
YSI™
60510-10 |
393.7 in. | 10 m |
368.00 €
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
12983487
|
YSI™
60510-30 |
1181.1 in. | 30 m |
569.00 €
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
12973487
|
YSI™
60510-20 |
787.4 in. | 20 m |
441.00 €
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||
Description
Conductivity Probes and Multiparameter Cables. Order other probes separately.
- The 60510 ISE (pH, ORP, pH/ORP, ammonium, nitrate, chloride) cables ONLY work with the YSI Pro Plus handheld Any of the ISE sensors can be connected to this cable at any time
Specifications
Cable management kit |