Learn More
Jenway™ 6320 Benchtop Visible Spectrophotometer
An easy-to-use instrument with a compact footprint.
Brand:  Jenway™ 99968-69
Product Code. 12421340
  • 4390.00 € / Each
Explore more special offers
Description
Description
  • Recalls previous settings at power up
  • Analog and RS-232 outputs let you interface with printers and PCs
  • Dual-line LCD shows error messages, prompts, mode indication, and concentration units
Specifications
Specifications
Xenon Flash
Education, quality control, environmental and clinical analysis
Independently housed and isolated with lenses
8 nm, 6 nm over the UV range
Mains lead, Pk. 100 disposable cuvettes, 10 x 10 mm cell holder, interface cable and PC application software on CD-ROM