Learn More
Masterflex™ 65mm Correlated PTFE Flowmeter
Brand:  Masterflex™ 32044-57
Product Code. 11735259
  • 459.00 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Variable Area Flowmeter
PTFE, Glass
1/8 in. NPT (F)
For high-purity applications with flexibility for a broad array of gases
65 mm
±2% (Full Scale)
100 psig
122 mL/min. CO2, 94 mL/min. O2, 113 mL/min. N2, 86 mL/min. Ar, 284 mL/min. H2
±0.25%
65°C